CAS RN 123-62-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Promotions Available
Propionic anhydride, 99
Propionic anhydride, 99
CAS: 123-62-6 Molecular Formula: C6;H1;0;O3; Molecular Weight (g/mol): 130.14
2
Propionic Anhydride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Propionic Anhydride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.