accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OCH3OH3CO

CAS RN 123-63-7

IUPAC Name: 2,4,6-trimethyl-1,3,5-trioxane
Synonyms: paral paraldehyde paraldehyd paracetaldehyde p-acetaldehyde acetaldehyde trimer acetaldehyde, trimer elaldehyde paraldeide paraacetaldehyde
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: SQYNKIJPMDEDEG-UHFFFAOYSA-N
SMILES: CC1OC(C)OC(C)O1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)