accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3NH2

CAS RN 123-82-0

IUPAC Name: heptan-2-amine
Synonyms: tuaminoheptane tuaminoheptano heptamine (sympathomimetic) tuaminoheptanum armeen L-7 tuaminheptanum tuaminoheptane (inn), its isomers and salts tuaminoheptane (substance)
Molecular Weight (g/mol): 115.22
Molecular Formula: C7H17N
InChi Key: VSRBKQFNFZQRBM-UHFFFAOYNA-N
SMILES: CCCCCC(C)N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)