accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3O

CAS RN 123-86-4

IUPAC Name: butyl acetate
Synonyms: butyl acetate butylester kyseliny octove butyle (acetate de) butylacetaten octan n-butylu n-butyl acetate (substance) N-butyl acetate essigsaeurebutylester acetate de butyle n-butyl acetate (natural)
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: DKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILES: CCCCOC(C)=O
Quantity 
  • (13)
  • (2)
  • (3)
  • (1)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
Boiling Point 
  • (9)
  • (14)
  • (4)
  • (6)
  • (1)
  • (11)
Percent Purity 
  • (2)
  • (1)
  • (8)
  • (4)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (26)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (10)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (13)
  • (9)
  • (1)
  • (13)
  • (2)
  • (3)
  • (1)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (9)
  • (14)
  • (4)
  • (6)
  • (1)
  • (11)
  • (2)
  • (1)
  • (8)
  • (4)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (26)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)