accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 123-95-5

IUPAC Name: butyl octadecanoate
Synonyms: butyl stearate N-butyl stearate
Molecular Weight (g/mol): 340.59
Molecular Formula: C22H44O2
InChi Key: ULBTUVJTXULMLP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCCCC
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results