CAS RN 123333-66-4
CAS RN 123333-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Potassium tellurite monohydrate, 97%
CAS: 123333-66-4 Molecular Formula: K2O3Te·H2O MDL Number: MFCD00149923 Synonym: potassium tellurite,potassium tellurate iv,dipotassium tellurite,dipotassium trioxotellurate,telluric acid, dipotassium salt,potassiium tellurite iv,telluric acid h2teo3 , dipotassium salt,dipotassium ion tellurite,acmc-1bm2e