accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 123333-71-1

IUPAC Name: hydrogen 2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrate chloride
Synonyms: hydrogen histidine hydrate chloride
Molecular Weight (g/mol): 209.63
Molecular Formula: C6H12ClN3O3
InChi Key: CMXXUDSWGMGYLZ-UHFFFAOYNA-N
SMILES: [H+].O.[Cl-].NC(CC1=CN=CN1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)