accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 123334-05-4

CH3SOOOCH3SOOOZn

CAS RN 123334-05-4

IUPAC Name: zinc(2+) bis(4-methylbenzene-1-sulfonate) hydrate
Synonyms: zinc(2+) hydrate ditosylate
Molecular Weight (g/mol): 425.78
Molecular Formula: C14H16O7S2Zn
InChi Key: JCOXFVWERPVZPB-UHFFFAOYSA-L
SMILES: O.[Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)