accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3PNCH3H3CCH3CH3H3CH3CH3CCH3PNCH3H3CCH3CH3H3CPd

CAS RN 1233717-68-4

IUPAC Name: bis(4-(di-tert-butylphosphanyl)-N,N-dimethylaniline) palladium
Synonyms: bis(4-(di-tert-butylphosphanyl)-N,N-dimethylaniline) palladium
Molecular Weight (g/mol): 637.18
Molecular Formula: C32H56N2P2Pd
InChi Key: SSPOQURGNAWORH-UHFFFAOYSA-N
SMILES: [Pd].CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results