CAS RN 123549-14-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-O-(p-Nitrophenyl)-Alpha-D-N-acetylneuraminic Acid, Sodium Salt, X Hydrate, TRC
CAS: 123549-14-4 Molecular Formula: C17 H21 N2 O11 . Na . H2 O Molecular Weight (g/mol): 470.36 IUPAC Name: sodium;(2S,4S,5R,6R)-5-acetamido-4-hydroxy-2-(4-nitrophenoxy)-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylate;hydrate SMILES: O.[Na+].CC(=O)N[C@@H]1[C@@H](O)C[C@@](Oc2ccc(cc2)[N+](=O)[O-])(O[C@H]1[C@H](O)[C@H](O)CO)C(=O)[O-]