accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrNFFFCl

CAS RN 123637-51-4

IUPAC Name: 6-(bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
Synonyms: 6-(bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
Molecular Weight (g/mol): 324.53
Molecular Formula: C11H6BrClF3N
InChi Key: SJBKLFYWXYFAGT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC2=CC=C(CBr)C=C2C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results