accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONH2F

CAS RN 123652-95-9

IUPAC Name: 1-(3-fluoro-4-methoxyphenyl)methanamine
Synonyms: 1-(3-fluoro-4-methoxyphenyl)methanamine
Molecular Weight (g/mol): 155.17
Molecular Formula: C8H10FNO
InChi Key: AZHAJNNLBSKSOB-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results