accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 123843-65-2

H3COFOHOF

CAS RN 123843-65-2

IUPAC Name: 2,6-difluoro-4-methoxybenzoic acid
Synonyms: 2,6-difluoro-4-methoxybenzoic acid
Molecular Weight (g/mol): 188.13
Molecular Formula: C8H6F2O3
InChi Key: LGHVYSAINQRZJQ-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(C(O)=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results