CAS RN 123855-55-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
p-Nitrophenyl 2-(Furfurylsulfinyl)acetate, TRC
CAS: 123855-55-0 Molecular Formula: C13H11NO6S Molecular Weight (g/mol): 309.29 Synonym: 2-[(2-Furanylmethyl)sulfinyl]acetic Acid 4-Nitrophenyl Ester,[(2-Furanylmethyl)sulfinyl]acetic Acid 4-Nitrophenyl Ester; IUPAC Name: (4-nitrophenyl) 2-(furan-2-ylmethylsulfinyl)acetate SMILES: [O-][N+](=O)c1ccc(OC(=O)CS(=O)Cc2occc2)cc1