accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrH3CNNH2H2N

CAS RN 1239-45-8

IUPAC Name: 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Synonyms: ethidium bromide EtBr 胡米溴铵 dromilac homidium bromide
Molecular Weight (g/mol): 394.32
Molecular Formula: C21H20BrN3
InChi Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Molecular Weight (g/mol) 
  • (2)
  • (22)
Quantity 
  • (6)
  • (1)
  • (4)
  • (4)
  • (7)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (4)
  • (2)
Grade 
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (10)
  • (3)
  • (8)
  • (2)
  • (2)
  • (22)
  • (6)
  • (1)
  • (4)
  • (4)
  • (7)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)