accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 123987-12-2

IUPAC Name: 4-[2-(hydroxymethyl)phenyl]-1-methylpiperazin-1-ium
Synonyms: 4-[2-(hydroxymethyl)phenyl]-1-methylpiperazin-1-ium
Molecular Weight (g/mol): 207.3
Molecular Formula: C12H19N2O
InChi Key: TWPYBKBPHCMUIS-UHFFFAOYSA-O
SMILES: C[NH+]1CCN(CC1)C1=CC=CC=C1CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results