CAS RN 124-02-7
IUPAC Name:
bis(prop-2-en-1-yl)amine
Synonyms:
diallylamine
Molecular Weight (g/mol):
97.16
Molecular Formula:
C6H11N
InChi Key:
DYUWTXWIYMHBQS-UHFFFAOYSA-N
SMILES:
C=CCNCC=C
1
–
1
of
1
results
1
Diallylamine, 97%
CAS: 124-02-7 Molecular Formula: C6H11N Molecular Weight (g/mol): 97.16 MDL Number: MFCD00008642 InChI Key: DYUWTXWIYMHBQS-UHFFFAOYSA-N Synonym: diallylamine,di-2-propenylamine,2-propen-1-amine, n-2-propenyl,amine, diallyl,bis prop-2-en-1-yl amine,n-2-propenyl-2-propen-1-amine,unii-n18exb6v6p,n,n-diallylamine,ccris 4776,n-allylprop-2-en-1-amine PubChem CID: 31279 IUPAC Name: N-prop-2-enylprop-2-en-1-amine SMILES: C=CCNCC=C