accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3NH2HN

CAS RN 124-42-5

IUPAC Name: hydrogen ethanimidamide chloride
Synonyms: hydrogen acetamidine chloride
Molecular Weight (g/mol): 94.54
Molecular Formula: C2H7ClN2
InChi Key: WCQOBLXWLRDEQA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(N)=N
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)