accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 124-43-6

IUPAC Name: peroxol; urea
Synonyms: hydrogen peroxide - urea
Molecular Weight (g/mol): 94.07
Molecular Formula: CH6N2O3
InChi Key: AQLJVWUFPCUVLO-UHFFFAOYSA-N
SMILES: OO.NC(N)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results