accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 124-63-0

IUPAC Name: methanesulfonyl chloride
Synonyms: methanesulfonyl chloride
Molecular Weight (g/mol): 114.54
Molecular Formula: CH3ClO2S
InChi Key: QARBMVPHQWIHKH-UHFFFAOYSA-N
SMILES: CS(Cl)(=O)=O
Boiling Point 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)