accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SFFFFFClONH2

CAS RN 1240257-35-5

IUPAC Name: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzamide
Synonyms: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzamide
Molecular Weight (g/mol): 281.63
Molecular Formula: C7H5ClF5NOS
InChi Key: SCYISRHUJQHNFD-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results