accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1240257-83-3

SFFFFFOH2N

CAS RN 1240257-83-3

IUPAC Name: 3-(pentafluoro-λ⁶-sulfanyl)benzamide
Synonyms: 3-(pentafluoro-λ⁶-sulfanyl)benzamide
Molecular Weight (g/mol): 247.18
Molecular Formula: C7H6F5NOS
InChi Key: YLMGGQIZDXTBKX-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(=CC=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results