accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1244855-59-1

NNNClI

CAS RN 1244855-59-1

IUPAC Name: 7-benzyl-4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 7-benzyl-4-chloro-5-iodopyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 369.59
Molecular Formula: C13H9ClIN3
InChi Key: NWGKGMFQFVMROS-UHFFFAOYSA-N
SMILES: ClC1=C2C(I)=CN(CC3=CC=CC=C3)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results