accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 1246467-58-2

IUPAC Name: 3,5-bis(pyren-1-yl)pyridine
Synonyms: 3,5-bis(pyren-1-yl)pyridine
Molecular Weight (g/mol): 479.58
Molecular Formula: C37H21N
InChi Key: YNTQHSDEZACXAP-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C3C(=CC=C4C=CC(=C1)C2=C34)C1=CN=CC(=C1)C1=C2C=CC3=CC=CC4=CC=C(C=C1)C2=C34
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)