accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 124811-71-8

NFFFOH

CAS RN 124811-71-8

IUPAC Name: 4-hydroxy-3-(trifluoromethyl)benzonitrile
Synonyms: 4-hydroxy-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H4F3NO
InChi Key: QXIFYONUKBXFTH-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results