accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNNNNOO(S)CH3H2NOOCH3NH2HCl

CAS RN 124832-27-5

IUPAC Name: hydrogen 2-[(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate chloride
Synonyms: hydrogen valaciclovir chloride
Molecular Weight (g/mol): 360.8
Molecular Formula: C13H21ClN6O4
InChi Key: ZCDDBUOENGJMLV-QRPNPIFTSA-N
SMILES: [H+].[Cl-].CC(C)[C@H](N)C(=O)OCCOCN1C=NC2=C1NC(N)=NC2=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results