accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3HOOOCH3H3COHCH3

CAS RN 125-20-2

IUPAC Name: 3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Synonyms: thymolphthalein
Molecular Weight (g/mol): 430.54
Molecular Formula: C28H30O4
InChi Key: LDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (12)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
  • (4)
  • (3)
Color 
  • (2)
  • (5)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (7)
  • (3)
  • (4)
  • (4)
  • (3)
  • (12)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (5)
  • (4)