accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 12501-23-4

IUPAC Name: phosphoric acid dodecakis(trioxotungsten) hydrate
Synonyms: phosphoric acid dodecakis(tungsten trioxide) hydrate
Molecular Weight (g/mol): 2898.05
Molecular Formula: H5O41PW12
InChi Key: AVFBYUADVDVJQL-UHFFFAOYSA-N
SMILES: O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)