accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 125110-14-7

IUPAC Name: (6R,7R)-7-[(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid trihydrate
Synonyms: cefixim trihydrate
Molecular Weight (g/mol): 507.49
Molecular Formula: C16H21N5O10S2
InChi Key: IPYWNMVPZOAFOQ-XZVQIPOZSA-N
SMILES: O.O.O.NC1=NC(=CS1)C(=N/OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)