accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCH3OOP

CAS RN 1253-47-0

IUPAC Name: {[4-(methoxycarbonyl)phenyl]methyl}triphenylphosphanium chloride
Synonyms: {[4-(methoxycarbonyl)phenyl]methyl}triphenylphosphanium chloride
Molecular Weight (g/mol): 446.91
Molecular Formula: C27H24ClO2P
InChi Key: NKKAHXNONUTMEY-UHFFFAOYSA-M
SMILES: [Cl-].COC(=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results