accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 125552-89-8

BrO

CAS RN 125552-89-8

IUPAC Name: 4-(bromomethyl)oxane
Synonyms: 4-(bromomethyl)oxane
Molecular Weight (g/mol): 179.06
Molecular Formula: C6H11BrO
InChi Key: LMOOYAKLEOGKJR-UHFFFAOYSA-N
SMILES: BrCC1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results