accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SNH3CCH3H3COH

CAS RN 1256483-61-0

IUPAC Name: (2-tert-butyl-4-methyl-1,3-thiazol-5-yl)methanol
Synonyms: (2-tert-butyl-4-methyl-1,3-thiazol-5-yl)methanol
Molecular Weight (g/mol): 185.29
Molecular Formula: C9H15NOS
InChi Key: HJESPMQKUUFUGH-UHFFFAOYSA-N
SMILES: CC1=C(CO)SC(=N1)C(C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results