accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNH2NNOHN(E)(E)

CAS RN 1256486-22-2

IUPAC Name: 1-[4-(5-methyl-2,3-dihydro-1H-1,3-benzodiazol-2-ylidene)cyclohexa-2,5-dien-1-ylidene]-1-nitrosomethanamine
Synonyms: 1-[4-(5-methyl-1,3-dihydro-1,3-benzodiazol-2-ylidene)cyclohexa-2,5-dien-1-ylidene]-1-nitrosomethanamine
Molecular Weight (g/mol): 266.3
Molecular Formula: C15H14N4O
InChi Key: SQIGGAWUCHCZKN-UHFFFAOYSA-N
SMILES: CC1=CC2=C(NC(N2)=C2C=CC(C=C2)=C(N)N=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results