accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNHHOHNHNNHNO(Z)(Z)

CAS RN 1256486-33-5

IUPAC Name: 2-{4-[amino(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-ylidene}-N-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboximidamide
Synonyms: 2-{4-[amino(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-ylidene}-N-oxo-1,3-dihydro-1,3-benzodiazole-5-carboximidamide
Molecular Weight (g/mol): 310.32
Molecular Formula: C15H14N6O2
InChi Key: GLMSCQQNOMUUFK-UHFFFAOYSA-N
SMILES: NC(NO)=C1C=CC(C=C1)=C1NC2=C(N1)C=C(C=C2)C(=N)N=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results