accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 125700-67-6

IUPAC Name: boron(3+) (1H-1,2,3-benzotriazol-1-yl)[bis(dimethylamino)methylidene]oxidanium tetrafluoride
Synonyms: boron(3+) HBTU tetrafluoride
Molecular Weight (g/mol): 321.09
Molecular Formula: C11H16BF4N5O
InChi Key: SHGLJDNRZIDILW-UHFFFAOYSA-J
SMILES: [B+3].[F-].[F-].[F-].[F-].CN(C)C(=[O+]N1N=NC2=CC=CC=C12)N(C)C
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results