accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNNNNNNH2OFeFe

CAS RN 12582-61-5

IUPAC Name: bis(λ²-iron(2+)) bis(2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide) hydrate
Synonyms: bis(λ²-iron(2+)) bis(2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide) hydrate
Molecular Weight (g/mol): 1355.18
Molecular Formula: C88H58Fe2N8O
InChi Key: JTFKJCUSDKAMIA-UHFFFAOYSA-N
SMILES: O.[Fe++].[Fe++].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1.[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results