accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFFOH

CAS RN 125903-81-3

IUPAC Name: [3-(difluoromethoxy)phenyl]methanol
Synonyms: [3-(difluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 174.15
Molecular Formula: C8H8F2O2
InChi Key: BBDUCPSYPRGPGO-UHFFFAOYSA-N
SMILES: OCC1=CC(OC(F)F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results