accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 125941-87-9

CH3NHOClHCl

CAS RN 125941-87-9

IUPAC Name: 8-chloro-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepin-7-ol hydrochloride
Synonyms: 8-chloro-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol hydrochloride
Molecular Weight (g/mol): 324.25
Molecular Formula: C17H19Cl2NO
InChi Key: OYCAEWMSOPMASE-UHFFFAOYNA-N
SMILES: Cl.CN1CCC2=CC(Cl)=C(O)C=C2C(C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results