accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 125995-03-1

H3CCH3N(R)(R)OHOOFONH

CAS RN 125995-03-1

IUPAC Name: 5-(4-fluorophenyl)-1-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-N,4-diphenyl-2-(propan-2-yl)-1H-pyrrole-3-carboxamide
Synonyms: 5-(4-fluorophenyl)-1-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-2-isopropyl-N,4-diphenylpyrrole-3-carboxamide
Molecular Weight (g/mol): 540.64
Molecular Formula: C33H33FN2O4
InChi Key: OUCSEDFVYPBLLF-KAYWLYCHSA-N
SMILES: CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H]1C[C@@H](O)CC(=O)O1)C1=CC=C(F)C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results