accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OO(R)O(R)O(S)OH3COO(R)OH3CO(R)OCH3O(S)OCH3O(R)OH3CO(S)OH3CO(R)OH3CO

CAS RN 126-14-7

IUPAC Name: [(2R,3R,4S,5S)-3,4-bis(acetyloxy)-5-[(acetyloxy)methyl]-5-{[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxolan-2-yl]methyl acetate
Synonyms: [(2R,3R,4S,5S)-3,4-bis(acetyloxy)-5-[(acetyloxy)methyl]-5-{[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxolan-2-yl]methyl acetate octaacetyl sucrose sucrose octaacetate sucrose octa acetate octaacetylsucrose octa-O-acetylsucrose
Molecular Weight (g/mol): 678.59
Molecular Formula: C28H38O19
InChi Key: ZIJKGAXBCRWEOL-SAXBRCJISA-N
SMILES: CC(=O)OC[C@H]1O[C@@](COC(C)=O)(O[C@H]2O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)