CAS RN 126-38-5
IUPAC Name:
1-bromo-2,2-dimethoxypropane
Synonyms:
1-bromo-2,2-dimethoxypropane
Molecular Weight (g/mol):
183.05
Molecular Formula:
C5H11BrO2
InChi Key:
SGTITUFGCGGICE-UHFFFAOYSA-N
SMILES:
COC(C)(CBr)OC
1
–
1
of
1
results
1
1-Bromo-2,2-dimethoxypropane, 98%
CAS: 126-38-5 Molecular Formula: C5H11BrO2 Molecular Weight (g/mol): 183.045 MDL Number: MFCD00000208 InChI Key: SGTITUFGCGGICE-UHFFFAOYSA-N Synonym: propane,1-bromo-2,2-dimethoxy,propane, 1-bromo-2,2-dimethoxy,bromoacetone dimethyl acetal,bromo-2,2dimethoxypropane,bromo-2,2-dimethoxypropane,1-bromo-2,2-dimethoxypropan,1-bromo-2,2-dimethoxyprpopane,1-bromo-2,2-dimethoxy propane,1-bromo-2,2-dimethoxy-propane,bromoacetone dimethyl ketal PubChem CID: 67175 IUPAC Name: 1-bromo-2,2-dimethoxypropane SMILES: CC(CBr)(OC)OC