CAS RN 126-71-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Phosphoric Acid Tris(2-methylpropyl) Ester, TRC
CAS: 126-71-6 Molecular Formula: C12 H27 O4 P Molecular Weight (g/mol): 266.31 Synonym: Phosphoric acid, tris(2-methylpropyl) ester,Isobutyl phosphate ((C4H9O)3PO) (6CI,7CI),Phosphoric acid, triisobutyl ester (8CI),Antifoam TIP,Daiguard 400,NSC 62222,Reomol TIBP,Triisobutyl phosphate IUPAC Name: tris(2-methylpropyl) phosphate SMILES: CC(C)COP(=O)(OCC(C)C)OCC(C)C
2
Phosphoric Acid Tris(2-methylpropyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.