accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH2CH2C

CAS RN 126-99-8

IUPAC Name: 2-chlorobuta-1,3-diene
Synonyms: chloroprene β-chloroprene chloropreen chloroprene (substance) chloroprene (stabilized) 2-cloro-1,3-butadiene ccris 873 2-chloor-1,3-butadieen C4H5cl cloroprene
Molecular Weight (g/mol): 88.53
Molecular Formula: C4H5Cl
InChi Key: YACLQRRMGMJLJV-UHFFFAOYSA-N
SMILES: ClC(=C)C=C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Solvent or Matrix 
  • (2)
  • (2)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results