accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HOOHOOOHOH(R)O(R)OH(S)OH(R)OH(R)HOOHO

CAS RN 1260-17-9

IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2R,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Synonyms: carminic acid cochineal extract san-ei gen san red 1 coccinellin sun red 1 carmine stain C.I. natural red 4 sanred 1 sun red no. 1 CI natural red 4
Molecular Weight (g/mol): 492.39
Molecular Formula: C22H20O13
InChi Key: DGQLVPJVXFOQEV-NGOCYOHBSA-N
SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C([C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C(O)C(O)=C1C2=O
Quantity 
  • (2)
  • (1)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results