accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 126120-85-2

IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxole-4-carboxylic acid
Synonyms: 2,2-difluoro-1,3-benzodioxole-4-carboxylic acid
Molecular Weight (g/mol): 202.11
Molecular Formula: C8H4F2O4
InChi Key: ZGAQVJDFFVTWJK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=C1OC(F)(F)O2
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results