accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHOOFFF

CAS RN 1261587-75-0

IUPAC Name: 3-[4-methoxy-3-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[4-methoxy-3-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 264.2
Molecular Formula: C11H11F3O4
InChi Key: DALLMZMEZXJMBE-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)(F)F)C=C(CCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results