accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOFFFOH2N

CAS RN 1261791-05-2

IUPAC Name: 4-chloro-2-(trifluoromethoxy)benzamide
Synonyms: 4-chloro-2-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 239.58
Molecular Formula: C8H5ClF3NO2
InChi Key: MTYIZCXOBKSDPF-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(OC(F)(F)F)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results