CAS RN 12619-68-0
CAS RN 12619-68-0
IUPAC Name:
tris(λ²-cobalt(2+)) diboron
Synonyms:
tris(λ²-cobalt(2+)) diboron
Molecular Weight (g/mol):
198.42
Molecular Formula:
B2Co3
InChi Key:
QSOMCIJHXYGMAO-UHFFFAOYSA-N
SMILES:
[B].[B].[Co++].[Co++].[Co++]
1
–
1
of
1
results
1
Cobalt boride, 99%, Thermo Scientific™
CAS: 12619-68-0 Molecular Formula: B2Co3+6 Molecular Weight (g/mol): 198.42 MDL Number: MFCD00146483 InChI Key: QSOMCIJHXYGMAO-UHFFFAOYSA-N Synonym: boron;cobalt 2+ PubChem CID: 121233614 IUPAC Name: boron;cobalt(2+) SMILES: [B].[B].[Co+2].[Co+2].[Co+2]