CAS RN 1262605-53-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyhalodiamide, TRC
CAS: 1262605-53-7 Molecular Formula: C22 H17 Cl F7 N3 O2 Molecular Weight (g/mol): 523.83 IUPAC Name: 3-chloro-2-N-(2-cyanopropan-2-yl)-1-N-[4-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)-2-methylphenyl]benzene-1,2-dicarboxamide SMILES: Cc1cc(ccc1NC(=O)c2cccc(Cl)c2C(=O)NC(C)(C)C#N)C(F)(C(F)(F)F)C(F)(F)F