accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1263-89-4

NH2(S)O(R)O(S)(R)HOO(R)OHO(R)(S)OH(R)NH2(S)H2NO(S)O(R)OH(S)HO(R)HO(R)H2N(R)NH2(R)OH(S)HOOHSOHOO

CAS RN 1263-89-4

IUPAC Name: (2S,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(2R,3S,4R)-5-{[(1R,3S,5R,6S)-3,5-diamino-2-{[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-hydroxycyclohexyl]oxy}-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Synonyms: paromomycin
Molecular Weight (g/mol): 713.71
Molecular Formula: C23H47N5O18S
InChi Key: LJRDOKAZOAKLDU-UMIPPVEZSA-N
SMILES: OS(O)(=O)=O.NC[C@@H]1O[C@H](O[C@@H]2[C@@H](CO)OC(O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)C3O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)[C@H](N)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Type 
  • (4)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)